siRNA Details
| Gene Symbol: VIM-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108060.1 | 4 | 390 | |||
| NR_108061.1 | 3 | 914 | ||||
| GenBank mRNA | BG492614.1 | - | 4 | 389 | ||
| BG495699.1 | - | 4 | 388 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

VIM-AS1| Gene Symbol: VIM-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108060.1 | 4 | 390 | |||
| NR_108061.1 | 3 | 914 | ||||
| GenBank mRNA | BG492614.1 | - | 4 | 389 | ||
| BG495699.1 | - | 4 | 388 | |||