siRNA Details
| Gene Symbol: LINC00242 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026781.1 | 1 | 77 | |||
| GenBank mRNA | AK056013.1 | - | 1 | 77 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000437615.1 | - | 1 | 77 | Chr. 6 - 169788790 - 169798825 [-] on Build GRCh38 | |

LINC00242
Genomic Map