siRNA Details
| Gene Symbol: LINC00582 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034037.1 | 1, 2 | 88 | |||
| GenBank mRNA | AI820788.1 | - | 1, 2 | 88 | ||
| BX092370.1 | - | 1, 2 | 88 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000448058.1 | - | 1, 2 | 88 | Chr. 1 - 231591292 - 231612090 [-] on Build GRCh38 | |

LINC00582
Genomic Map