siRNA Details
| Gene Symbol: LINC00710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015413.1 | 6 | 1053 | |||
| NR_024410.1 | 5 | 983 | ||||
| NR_024411.1 | 4 | 872 | ||||
| GenBank mRNA | BC033338.1 | - | 6 | 985 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00710| Gene Symbol: LINC00710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015413.1 | 6 | 1053 | |||
| NR_024410.1 | 5 | 983 | ||||
| NR_024411.1 | 4 | 872 | ||||
| GenBank mRNA | BC033338.1 | - | 6 | 985 | ||