siRNA Details
| Gene Symbol: LOC101928103 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110292.1 | 4 | 1203 | |||
| GenBank mRNA | AK056813.1 | - | 4 | 1203 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000607412.1 | - | 4 | 1203 | Chr. 2 - 214810229 - 214963274 [+] on Build GRCh38 | |

LOC101928103
Genomic Map