siRNA Details
| Gene Symbol: LOC102031319 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104163.1 | 2 | 134 | |||
| GenBank mRNA | BX107541.1 | - | 2 | 134 | ||
| BX118505.1 | - | 2 | 134 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000417447.1 | - | 2 | 101 | Chr. 10 - 32347397 - 32374488 [+] on Build GRCh38 | |

LOC102031319
Genomic Map