siRNA Details
Gene Symbol: LOC101927000 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132384.1 | 2 | 78 | |||
GenBank mRNA | BX118161.1 | - | 2 | 78 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000453149.1 | - | 2 | 78 | Chr. 7 - 561958 - 565619 [+] on Build GRCh38 |
