siRNA Details
| Gene Symbol: LOC101927000 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132384.1 | 2 | 266 | |||
| GenBank mRNA | BX118161.1 | - | 2 | 266 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000453149.1 | - | 2 | 266 | Chr. 7 - 561958 - 565619 [+] on Build GRCh38 | |

LOC101927000
Genomic Map