siRNA Details
Gene Symbol: LINC00858 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038220.1 | 5 | 1248 | |||
GenBank mRNA | AK056561.1 | - | 5 | 1248 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000415469.5 | - | 5 | 1248 | Chr. 10 - 84279980 - 84294659 [+] on Build GRCh38 |
