siRNA Details
| Gene Symbol: LINC00433 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047021.1 | 1 | 291 | |||
| GenBank mRNA | BI520070.1 | - | 1 | 297 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000424165.1 | - | 1 | 256 | Chr. 13 - 88540864 - 88542506 [+] on Build GRCh38 | |

LINC00433
Genomic Map