siRNA Details
Gene Symbol: LOC101929592 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125986.1 | 1 | 24 | |||
GenBank mRNA | BX280555.1 | - | 1 | 24 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000437753.1 | - | 1 | 24 | Chr. 1 - 43709392 - 43727343 [-] on Build GRCh38 |
