siRNA Details
Gene Symbol: LINC00362 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX107685.1 | - | 2 | 255 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000414345.2 | - | 2 | 222 | Chr. 13 - 23169835 - 23170597 [-] on Build GRCh38 |
