siRNA Details
| Gene Symbol: LINC00362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX107685.1 | - | 2 | 108 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000414345.2 | - | 2 | 75 | Chr. 13 - 23169835 - 23170597 [-] on Build GRCh38 | |

LINC00362
Genomic Map