siRNA Details
| Gene Symbol: LOC101927948 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110287.1 | 2 | 85 | |||
| GenBank mRNA | BC042517.1 | - | 2 | 85 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000439259.1 | - | 2 | 85 | Chr. 2 - 78088730 - 78127806 [+] on Build GRCh38 | |

LOC101927948
Genomic Map