siRNA Details
| Gene Symbol: LOC284950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038888.1 | 3 | 641 | |||
| GenBank mRNA | AK095038.1 | - | 3 | 642 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000366278.2 | - | 641 | Chr. 4 - 82898639 - 82899008 [-] on Build GRCh38 | ||

LOC284950
Genomic Map