siRNA Details
Gene Symbol: LINC01523 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109942.1 | 2 | 449 | |||
GenBank mRNA | BX108395.1 | - | 2 | 449 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000425385.1 | - | 2 | 271 | Chr. 20 - 47983404 - 47984313 [+] on Build GRCh38 |
