siRNA Details
| Gene Symbol: LOC101928696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121196.1 | 2 | 120 | |||
| GenBank mRNA | BX102231.1 | - | 2 | 120 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928696| Gene Symbol: LOC101928696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121196.1 | 2 | 120 | |||
| GenBank mRNA | BX102231.1 | - | 2 | 120 | ||