siRNA Details
Gene Symbol: LINC00379 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047004.1 | 4 | 554 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000427717.1 | - | 2 | 418 | Chr. 13 - 91127613 - 91131407 [-] on Build GRCh38 |
