siRNA Details
| Gene Symbol: LINC00379 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047004.1 | 4 | 448 | |||
| GenBank mRNA | CD387405.1 | - | 4 | 468 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000427717.1 | - | 2 | 312 | Chr. 13 - 91127613 - 91131407 [-] on Build GRCh38 | |

LINC00379
Genomic Map