siRNA Details
| Gene Symbol: LINC01204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104645.1 | 3 | 1239 | |||
| GenBank mRNA | BC040858.1 | - | 3 | 1240 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000435394.5 | - | 3 | 1239 | Chr. X - 45505388 - 45523644 [+] on Build GRCh38 | |

LINC01204
Genomic Map