siRNA Details
| Gene Symbol: LINC01204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104644.1 | 4 | 960 | |||
| GenBank mRNA | AK127697.1 | - | 4 | 936 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000609127.1 | - | 4 | 936 | Chr. X - 45505412 - 45527239 [+] on Build GRCh38 | |

LINC01204
Genomic Map