siRNA Details
Gene Symbol: LINC01204 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104644.1 | 4 | 1866 | |||
GenBank mRNA | AK127697.1 | - | 4 | 1842 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000609127.1 | - | 4 | 1842 | Chr. X - 45505412 - 45527239 [+] on Build GRCh38 |
