siRNA Details
| Gene Symbol: LOC51145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122032.1 | 1 | 249 | |||
| GenBank mRNA | AA397744.1 | - | 1 | 249 | ||
| AF159054.1 | - | 1 | 872 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC51145| Gene Symbol: LOC51145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122032.1 | 1 | 249 | |||
| GenBank mRNA | AA397744.1 | - | 1 | 249 | ||
| AF159054.1 | - | 1 | 872 | |||