siRNA Details
| Gene Symbol: LINC01079 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BI715219.1 | - | 2 | 325 | ||
| BM503984.1 | - | 2 | 319 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000431171.1 | - | 2 | 325 | Chr. 13 - 27373348 - 27374366 [+] on Build GRCh38 | |

LINC01079
Genomic Map