siRNA Details
| Gene Symbol: LOC101927419 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120621.1 | 3 | 533 | |||
| GenBank mRNA | AK092205.1 | - | 3 | 533 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927419| Gene Symbol: LOC101927419 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120621.1 | 3 | 533 | |||
| GenBank mRNA | AK092205.1 | - | 3 | 533 | ||