siRNA Details
| Gene Symbol: LOC101928794 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_111908.1 | 2 | 727 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000451496.1 | - | 2 | 526 | Chr. 5 - 67001383 - 67003953 [-] on Build GRCh38 | |

LOC101928794
Genomic Map