siRNA Details
| Gene Symbol: LINC01150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120534.1 | 3 | 570 | |||
| GenBank mRNA | L38282.1 | - | 2 | 131 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01150| Gene Symbol: LINC01150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120534.1 | 3 | 570 | |||
| GenBank mRNA | L38282.1 | - | 2 | 131 | ||