siRNA Details
Gene Symbol: MIR194-2HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_133638.1 | 2 | 2359 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000413053.2 | - | 2359 | Chr. 11 - 64891426 - 64893449 [-] on Build GRCh38 |
