siRNA Details
| Gene Symbol: LOC101929460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134622.1 | 2 | 910 | |||
| GenBank mRNA | BC040917.1 | - | 2 | 910 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000419800.1 | - | 2 | 909 | Chr. 6 - 169034197 - 169035642 [-] on Build GRCh38 | |

LOC101929460
Genomic Map