siRNA Details
Gene Symbol: LINC01159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110373.1 | 2 | 2713 | |||
GenBank mRNA | AK095498.1 | - | 2 | 2501 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000433433.1 | - | 2 | 2570 | Chr. 2 - 104865497 - 104872452 [-] on Build GRCh38 |
