siRNA Details
| Gene Symbol: LINC01159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110373.1 | 2 | 1987 | |||
| GenBank mRNA | AK095498.1 | - | 2 | 1775 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000433433.1 | - | 2 | 1844 | Chr. 2 - 104865497 - 104872452 [-] on Build GRCh38 | |

LINC01159
Genomic Map