siRNA Details
| Gene Symbol: LOC105378470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134331.1 | 2 | 629 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000424701.1 | - | 2 | 503 | Chr. 10 - 106140291 - 106158145 [+] on Build GRCh38 | |

LOC105378470
Genomic Map