siRNA Details
Gene Symbol: LOC101927165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135590.1 | 2 | 657 | |||
GenBank mRNA | BC040311.1 | - | 2 | 654 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000451034.1 | - | 2 | 657 | Chr. 2 - 56077417 - 56090382 [-] on Build GRCh38 |
