siRNA Details
| Gene Symbol: KIF25-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103750.1 | 2 | 321 | |||
| GenBank mRNA | AB016899.1 | - | 1 | 625 | ||
| AB016900.1 | - | 2 | 321 | |||
| BC112215.1 | - | 1 | 56 | |||
| BC143766.1 | - | 1 | 56 | 9052280 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIF25-AS1| Gene Symbol: KIF25-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103750.1 | 2 | 321 | |||
| GenBank mRNA | AB016899.1 | - | 1 | 625 | ||
| AB016900.1 | - | 2 | 321 | |||
| BC112215.1 | - | 1 | 56 | |||
| BC143766.1 | - | 1 | 56 | 9052280 | ||