siRNA Details
Gene Symbol: KIF25-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103750.1 | 4 | 1267 | |||
GenBank mRNA | AB016900.1 | - | 4 | 1267 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000414364.1 | - | 4 | 1267 | Chr. 6 - 167994172 - 167997077 [-] on Build GRCh38 |
