siRNA Details
| Gene Symbol: LOC101928433 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136240.1 | 3 | 281 | |||
| GenBank mRNA | DB477374.1 | - | 3 | 282 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000450930.1 | - | 2 | 459 | Chr. 6 - 16761912 - 16762618 [+] on Build GRCh38 | |

LOC101928433
Genomic Map