siRNA Details
| Gene Symbol: FLJ31356 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103831.1 | 2 | 1837 | |||
| GenBank mRNA | AK055918.1 | - | 2 | 1837 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000427929.5 | - | 2 | 1837 | Chr. 2 - 28384409 - 28394672 [-] on Build GRCh38 | |

FLJ31356
Genomic Map