siRNA Details
| Gene Symbol: LOC105373368 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136578.1 | 2 | 94 | |||
| GenBank mRNA | BG182954.1 | - | 1 | 51 | ||
| BQ081842.1 | - | 2 | 210 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000445330.1 | - | 2 | 210 | Chr. X - 151904432 - 151911199 [-] on Build GRCh38 | |

LOC105373368
Genomic Map