siRNA Details
| Gene Symbol: LINC00709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108039.1 | 2 | 378 | |||
| GenBank mRNA | BX111793.1 | - | 2 | 378 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00709| Gene Symbol: LINC00709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108039.1 | 2 | 378 | |||
| GenBank mRNA | BX111793.1 | - | 2 | 378 | ||