siRNA Details
| Gene Symbol: LOC284632 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027087.2 | 3 | 833 | |||
| GenBank mRNA | BC033556.1 | - | 3 | 833 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000439059.2 | - | 3 | 833 | Chr. 1 - 24200240 - 24211693 [+] on Build GRCh38 | |

LOC284632
Genomic Map