siRNA Details
| Gene Symbol: TRPM2-AS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109964.1 | 3 | 851 | |||
| GenBank mRNA | AK057275.1 | - | 2 | 2036 | ||
| EU362988.1 | - | 3 | 851 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRPM2-AS| Gene Symbol: TRPM2-AS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109964.1 | 3 | 851 | |||
| GenBank mRNA | AK057275.1 | - | 2 | 2036 | ||
| EU362988.1 | - | 3 | 851 | |||