siRNA Details
| Gene Symbol: CDC42-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF086337.1 | - | 2 | 545 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000431803.2 | - | 2 | 517 | Chr. 1 - 22059197 - 22064199 [+] on Build GRCh38 | |

CDC42-IT1
Genomic Map