siRNA Details
| Gene Symbol: TCERG1L-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120623.1 | 2 | 399 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000436942.1 | - | 2 | 399 | Chr. 10 - 131095218 - 131095777 [+] on Build GRCh38 | |

TCERG1L-AS1
Genomic Map