siRNA Details
| Gene Symbol: LOC285389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136181.1 | 4 | 2435 | |||
| GenBank mRNA | BC038368.1 | - | 4 | 2435 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000432154.5 | - | 4 | 2435 | Chr. 3 - 194048920 - 194069529 [-] on Build GRCh38 | |

LOC285389
Genomic Map