siRNA Details
Gene Symbol: LOC285389 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136181.1 | 3 | 1722 | |||
GenBank mRNA | BC038368.1 | - | 3 | 1722 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000432154.5 | - | 3 | 1722 | Chr. 3 - 194048920 - 194069529 [-] on Build GRCh38 |
