siRNA Details
| Gene Symbol: LINC01433 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033917.1 | 3 | 646 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000419003.2 | - | 3 | 671 | Chr. 20 - 4193065 - 4195943 [+] on Build GRCh38 | |

LINC01433
Genomic Map