siRNA Details
| Gene Symbol: LOC105375556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133929.1 | 1 | 311 | |||
| GenBank mRNA | BF188822.1 | - | 1 | 296 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105375556| Gene Symbol: LOC105375556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133929.1 | 1 | 311 | |||
| GenBank mRNA | BF188822.1 | - | 1 | 296 | ||