siRNA Details
Gene Symbol: MIRLET7DHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046163.1 | 4 | 536 | |||
NR_046165.1 | 3 | 477 | ||||
GenBank mRNA | BC036695.1 | - | 4 | 515 | ||
BC045813.1 | - | 3 | 477 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MIRLET7DHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046163.1 | 4 | 536 | |||
NR_046165.1 | 3 | 477 | ||||
GenBank mRNA | BC036695.1 | - | 4 | 515 | ||
BC045813.1 | - | 3 | 477 |