siRNA Details
Gene Symbol: NAALADL2-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046390.1 | 3 | 136 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000436929.1 | - | 2 | 83 | Chr. 3 - 175079312 - 175112600 [-] on Build GRCh38 |
