siRNA Details
| Gene Symbol: LOC100505664 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK091765.1 | - | 1 | 2285 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000411595.2 | - | 652 | Chr. 20 - 23188128 - 23190307 [+] on Build GRCh38 | ||

LOC100505664
Genomic Map